Skip to Content
Merck
All Photos(1)

Key Documents

407984

Sigma-Aldrich

1,1,3-Tris(2-methyl-4-hydroxy-5-tert-butylphenyl)butane

Synonym(s):

4,4′,4′′-(1-Methyl-1-propanyl-3-ylidene)tris[2-(1,1-dimethylethyl)-5-methylphenol]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H52O3
CAS Number:
Molecular Weight:
544.81
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mp

183-190 °C (lit.)

SMILES string

CC(CC(c1cc(c(O)cc1C)C(C)(C)C)c2cc(c(O)cc2C)C(C)(C)C)c3cc(c(O)cc3C)C(C)(C)C

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sibel Ügdüler et al.
Waste management (New York, N.Y.), 104, 148-182 (2020-01-25)
Additives are ubiquitously used in plastics to improve their functionality. However, they are not always desirable in their 'second life' and are a major bottleneck for chemical recycling. Although research on extraction techniques for efficient removal of additives is increasing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service