Skip to Content
Merck
All Photos(1)

Documents

357723

Sigma-Aldrich

7-Methoxy-2-benzofurancarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

221-223 °C (lit.)

SMILES string

COc1cccc2cc(oc12)C(O)=O

InChI

1S/C10H8O4/c1-13-7-4-2-3-6-5-8(10(11)12)14-9(6)7/h2-5H,1H3,(H,11,12)

InChI key

UOCNTRAAJNWDND-UHFFFAOYSA-N

Application

7-Methoxy-2-benzofurancarboxylic acid may be used in the synthesis of fluorous counterpart of the Marshall resin, 4-(1H,1H,2H,2H-perfluorodecylsulfanyl)phenol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Hiu-Tung Chen et al.
Organic letters, 5(7), 1015-1017 (2003-03-28)
[structure: see text] The fluorous counterpart of the Marshall resin, 4-(1H,1H,2H,2H-perfluorodecylsulfanyl)phenol (FluoMar), is prepared by S-alkylation of 4-mercaptophenol with C(8)F(17)CH(2)CH(2)I and employed in the synthesis of amide and diamide analogues. The final products are purified by solid-phase extraction (SPE) over

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service