Skip to Content
Merck
All Photos(1)

Documents

336858

Sigma-Aldrich

Dichloroethylphosphine

95%

Synonym(s):

Ethyldichlorophosphine, Ethylphosphorous dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5PCl2
CAS Number:
Molecular Weight:
130.94
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

95%

form

liquid

reaction suitability

reagent type: ligand

refractive index

n20/D 1.495 (lit.)

bp

112 °C (lit.)

density

1.26 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CCP(Cl)Cl

InChI

1S/C2H5Cl2P/c1-2-5(3)4/h2H2,1H3

InChI key

JHNJGLVSPIMBLD-UHFFFAOYSA-N

Application

Catalyst in palladium-catalyzed Suzuki-Miyaura cross-coupling

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Pyr. Liq. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service