232831
Ethylamine hydrochloride
98%
Synonym(s):
N-Ethylammonium chloride, Ethylammonium chloride, Monoethylammonium chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
form
solid
mp
107-108 °C (lit.)
SMILES string
Cl[H].CCN
InChI
1S/C2H7N.ClH/c1-2-3;/h2-3H2,1H3;1H
InChI key
XWBDWHCCBGMXKG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Ethylamine hydrochloride can be used as a reactant to synthesize:
- Amino cellulose via reductive amination.
- N-triethylborazine by reacting with metal tetrahydroborates.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mechanochemical synthesis of N-triethylborazine
Russian Journal of Inorganic Chemistry, 54, 219-222 (2009)
Structural dynamics (Melville, N.Y.), 6(6), 064703-064703 (2019-12-24)
Deep eutectic solvents (DESs) prepared by mixing hydrogen-bond donor and acceptor molecules have been found to be of use in several applications. Recently, it was shown that DESs can enhance the peroxidation activity of cytochrome c. Here, to elucidate the
Amino-modified cellulose nanocrystals with adjustable hydrophobicity from combined regioselective oxidation and reductive amination
Carbohydrate Polymers, 136 (2016)
Molecules (Basel, Switzerland), 25(8) (2020-04-25)
The thermal, physical, and molecular dynamics of layered hybrid type (C2H5NH3)2MCl4 (M = 59Co, 63Cu, 65Zn, and 113Cd) crystals were investigated by thermogravimetric analysis (TGA) and magic angle spinning nuclear magnetic resonance (MAS NMR) spectroscopy. The temperatures of the onset
International journal of molecular sciences, 20(18) (2019-09-20)
A series of N-substituted poly(Gly-alter-Val) peptides were successfully synthesized for the systematic evaluation of the micellization behavior of alternating peptides. Three-component polymerization employing an aldehyde, a primary ammonium chloride, and potassium isocyanoacetate afforded four alternating peptides in excellent yields. We
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service