Skip to Content
Merck
All Photos(1)

Documents

203955

Sigma-Aldrich

trans-Diamminedichloropalladium(II)

99.998%

Synonym(s):

trans-Bisamminedichloropalladium, Transpalladin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(NH3)2Cl2
CAS Number:
Molecular Weight:
211.39
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.998%

reaction suitability

reagent type: catalyst
core: palladium

density

2.5 g/mL at 25 °C (lit.)

SMILES string

N.N.Cl[Pd]Cl

InChI

1S/2ClH.2H3N.Pd/h2*1H;2*1H3;/q;;;;+2/p-2

InChI key

WIEZTXFTOIBIOC-UHFFFAOYSA-L

Application

Catalyst for:
  • Stereoselective alkylation of aldol reactions
  • Solid phase oligosaccharide synthesis of N-glycans
  • Isomerization and cleavage of allyl ethers of carbohydrates

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service