Skip to Content
Merck
All Photos(1)

Documents

155764

Sigma-Aldrich

o-Phenylene phosphorochloridite

97%

Synonym(s):

2-Chloro-1,3,2-benzodioxaphosphole, o-Phenylene chlorophosphite, Chloro(1,2-phenylenedioxy)phosphine, Cyclic o-phenylene phosphorochloridite, Phosphorochloridous acid orthophenylene ester, Pyrocatechol phosphoryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClO2P
CAS Number:
Molecular Weight:
174.52
Beilstein:
135455
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

refractive index

n20/D 1.571 (lit.)

bp

80 °C/20 mmHg (lit.)

mp

30-35 °C (lit.)

density

1.466 g/mL at 25 °C (lit.)

SMILES string

ClP1Oc2ccccc2O1

InChI

1S/C6H4ClO2P/c7-10-8-5-3-1-2-4-6(5)9-10/h1-4H

InChI key

YUJYEGDMJZHLMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Catalyst in synthesis of bakkane sesquiterpenes
  • Reactant in reversible halogen-halogen ligand substitution at room temperature, and in irreversible halogen-O substitution at higher temperatures
  • Reactant in preparation of gold(I) trimethoxybenzonitrile phosphine isolated precatalysts
  • Reactant for preparation of pyrroles by TMSOTf-catalyzed Arbuzov reaction
  • Phosphorylation agent
  • Reactant for reparation of vinyl-substituted spirophosphoranes and vinylphosphonates via Markovnikov addition

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service