Skip to Content
Merck
All Photos(1)

Key Documents

ADVH7F3828E2

AChemBlock

((3aR,4R,6R,6aR)-6-(2,6-diamino-9H-purin-9-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methanol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 182.48

Vendor SKU: Q69447-100MG

CHF 182.48


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 182.48

About This Item

CAS Number:
Molecular Weight:
322.32
MDL number:

Vendor SKU: Q69447-100MG

CHF 182.48


Ships in 21 days - from Aldrich Partner.

Iupac Name

((3aR,4R,6R,6aR)-6-(2,6-diamino-9H-purin-9-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methanol

Inchi Code

1S/C13H18N6O4/c1-13(2)22-7-5(3-20)21-11(8(7)23-13)19-4-16-6-9(14)17-12(15)18-10(6)19/h4-5,7-8,11,20H,3H2,1-2H3,(H4,14,15,17,18)/t5-,7-,8-,11-/m1/s1

InChI key

GHTKDSBSFMMOKE-IOSLPCCCSA-N

storage temp.

Store at 0-8 C

Purity

0.95

Country of Origin

CN

Product Link

https://www.achemblock.com/q69447-3ar-4r-6r-6ar-6-2-6-diamino-9h-purin-9-yl-2-2-dimethyltetrahydrofuro-3-4-d-1-3-dioxol-4-yl-methanol.html

Synonym

((3ar,4r,6r,6ar)-6-(2,6-diamino-9h-purin-9-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methanol

Pictograms

Exclamation mark

Signal Word

warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/q69447-3ar-4r-6r-6ar-6-2-6-diamino-9h-purin-9-yl-2-2-dimethyltetrahydrofuro-3-4-d-1-3-dioxol-4-yl-methanol.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service