Skip to Content
Merck
All Photos(1)

Key Documents

ADVH14DBE282

AChemBlock

4,4-difluoro-1-phenylcyclohexan-1-amine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 268.21
1 G
CHF 592.77

Vendor SKU: P46725-250MG

CHF 268.21


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 268.21
1 G
CHF 592.77

About This Item

CAS Number:
Molecular Weight:
211.25
MDL number:

Vendor SKU: P46725-250MG

CHF 268.21


Ships in 21 days - from Aldrich Partner.

Iupac Name

4,4-difluoro-1-phenylcyclohexan-1-amine

Inchi Code

1S/C12H15F2N/c13-12(14)8-6-11(15,7-9-12)10-4-2-1-3-5-10/h1-5H,6-9,15H2

InChI key

XZQMJXOUHSANAK-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

97%

Country of Origin

CN

Product Link

https://www.achemblock.com/p46725-4-4-difluoro-1-phenylcyclohexan-1-amine.html

Synonym

4,4-difluoro-1-phenylcyclohexan-1-amine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.achemblock.com/p46725-4-4-difluoro-1-phenylcyclohexan-1-amine.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service