Skip to Content
Merck
All Photos(1)

Key Documents

AC5H5815FBA3

Accela ChemBio Inc.

2-Bromo-5-(4H-1,2,4-triazol-4-yl)pyrazine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 978.56

Vendor SKU: SY257810-1G

CHF 978.56


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
CHF 978.56

About This Item

CAS Number:
Molecular Weight:
226.04
MDL number:

Vendor SKU: SY257810-1G

CHF 978.56


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

2-bromo-5-(4H-1,2,4-triazol-4-yl)pyrazine

Inchi Code

1S/C6H4BrN5/c7-5-1-9-6(2-8-5)12-3-10-11-4-12/h1-4H

InChI key

XEHCPJMUIPSGGY-UHFFFAOYSA-N

storage temp.

+4C

Purity

98%

Country of Origin

CN

Shipping Temperature

RT

Physical Form

White to tan solid

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY257810&attname=GoodsCode

MSDS

http://www.accelachem.com/cn/msds/SY257810.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service