Skip to Content
Merck
All Photos(1)

Key Documents

AABH97CE8E5D

AA BLOCKS, INC.

1-[(1,3,5-trimethyl-1H-pyrazol-4-yl)sulfonyl]piperidine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 713.48
1 G
CHF 773.02
5 G
CHF 1’465.27

Vendor SKU: AA018WJK-500MG

CHF 713.48


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
500 MG
CHF 713.48
1 G
CHF 773.02
5 G
CHF 1’465.27

About This Item

CAS Number:
Molecular Weight:
301.37
MDL number:

Vendor SKU: AA018WJK-500MG

CHF 713.48


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

1-[(1,3,5-trimethyl-1H-pyrazol-4-yl)sulfonyl]-3-piperidinecarboxylic acid

Inchi Code

1S/C12H19N3O4S/c1-8-11(9(2)14(3)13-8)20(18,19)15-6-4-5-10(7-15)12(16)17/h10H,4-7H2,1-3H3,(H,16,17)

InChI key

CJYNCJMQAGDWKJ-UHFFFAOYSA-N

storage temp.

2~8 C

Purity

95%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/899703-32-3

Synonym

1-[(1,3,5-Trimethyl-1H-pyrazol-4-yl)sulfonyl]piperidine-3-carboxylicacid

MSDS

https://www.aablocks.com/downloadSds?cas=899703-32-3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service