Skip to Content
Merck
All Photos(1)

Key Documents

AABH9616BD9B

AA BLOCKS, INC.

5H-Benzo[a]phenoxazin-5-one, 9-(diethylamino)-2-hydroxy-

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 626.26
250 MG
CHF 941.93
1 G
CHF 2’284.89

Vendor SKU: AA002H4O-100MG

CHF 626.26


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 626.26
250 MG
CHF 941.93
1 G
CHF 2’284.89

About This Item

CAS Number:
Molecular Weight:
334.37
MDL number:

Vendor SKU: AA002H4O-100MG

CHF 626.26


Ships in 14 days - from Aldrich Partner.

Iupac Name

9-(diethylamino)-2-hydroxy-5H-benzo[a]phenoxazin-5-one

Inchi Code

1S/C20H18N2O3/c1-3-22(4-2)12-5-8-16-18(9-12)25-19-11-17(24)14-7-6-13(23)10-15(14)20(19)21-16/h5-11,23H,3-4H2,1-2H3

InChI key

BQNBSFWYKVAWQF-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/188712-75-6

Synonym

5H-Benzo[a]phenoxazin-5-one,9-(diethylamino)-2-hydroxy-

MSDS

https://www.aablocks.com/downloadSds?cas=188712-75-6


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service