Skip to Content
Merck
All Photos(1)

Key Documents

AABH3036E5B6

AA BLOCKS, INC.

5-amino-4-oxopentanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 475.35
5 G
CHF 1’167.60

Vendor SKU: AA00HANV-1G

CHF 475.35


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 475.35
5 G
CHF 1’167.60

About This Item

CAS Number:
Molecular Weight:
131.13
MDL number:

Vendor SKU: AA00HANV-1G

CHF 475.35


Ships in 14 days - from Aldrich Partner.

Iupac Name

5-amino-4-oxopentanoic acid

Inchi Code

1S/C5H9NO3/c6-3-4(7)1-2-5(8)9/h1-3,6H2,(H,8,9)

InChI key

ZGXJTSGNIOSYLO-UHFFFAOYSA-N

storage temp.

RT

Country of Origin

US

Product Link

https://www.aablocks.com/prod/106-60-5

Synonym

5-Amino-4-oxopentanoic acid

MSDS

https://www.aablocks.com/downloadSds?cas=106-60-5


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service