Skip to Content
Merck
All Photos(1)

Key Documents

R0260

Sigma-Aldrich

BamH I from Bacillus amyloliquefaciens H

Restriction Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

grade

Molecular Biology
for molecular biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Specificity

Recognition sequence: 5′-G/GATCC-3′
Cutting results: a 2-10-fold Bam HI overdigestion of 1 μg λ DNA substrate results in 100% cutting
Heat inactivation: 60 °C for 15 minutes.
Star activity: To prevent star activity, avoid suboptimal reaction conditions containing low salt concentration, high glycerol (>5%) and high pH 8.0.

Application

BamHI is a DNA restriction endonuclease that is used in molecular biology applications to cleave DNA at the recognition sequence 5′-G/GATCC-3′ to generate 5′-cohesive termini.

Other Notes

Supplied with 10x Restriction Enzyme Buffer SB (B8781).

Physical form

Solution in 10 mM Tris-HCl, pH 7.4 , 1 mM EDTA, 1mM dithioerythritol, 300 mM KCl, 0.01% Polydocanol (v/v), 50% glycerol (v/v), at 4°C

incubation buffer

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Manivasakam et al.
Nucleic acids research, 29(23), 4826-4833 (2001-12-01)
Mammalian cells repair DNA double-strand breaks by illegitimate end-joining or by homologous recombination. We investigated the effects of restriction enzymes on illegitimate and homologous DNA integration in mammalian cells. A plasmid containing the neo(R) expression cassette, which confers G418 resistance
J H Ellis et al.
Journal of immunology (Baltimore, Md. : 1950), 156(8), 2700-2709 (1996-04-15)
CD80 and CD86 are cell surface glycoproteins expressed on a variety of professional APCs. They have attracted much attention due to their function as potent costimulators of T lymphocyte function through their interaction with CD28 and possibly CTLA4. Because inhibitors
Isolation of a sequence-specific endonuclease (BamI) from Bacillus amyloliquefaciens H.
G A Wilson et al.
Journal of molecular biology, 97(1), 123-125 (1975-09-05)
C Kessler et al.
Gene, 92(1-2), 1-248 (1990-08-16)
The properties and sources of all known class-I, class-II and class-III restriction endonucleases (ENases) and DNA modification methyltransferases (MTases) are listed and newly subclassified according to their sequence specificity. In addition, the enzymes are distinguished in a novel manner according
Cong Zhu et al.
Nucleic acids research, 41(4), 2455-2465 (2013-01-11)
Zinc-finger nucleases (ZFNs) have been used for genome engineering in a wide variety of organisms; however, it remains challenging to design effective ZFNs for many genomic sequences using publicly available zinc-finger modules. This limitation is in part because of potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service