- Nortriptyline, a tricyclic antidepressant, inhibits voltage-dependent K
Nortriptyline, a tricyclic antidepressant, inhibits voltage-dependent K
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology (2017-03-11)
Sung Eun Shin, Hongliang Li, Han Sol Kim, Hye Won Kim, Mi Seon Seo, Kwon-Soo Ha, Eun-Taek Han, Seok-Ho Hong, Amy L Firth, Il-Whan Choi, Young Min Bae, Won Sun Park
PMID28280416
ZUSAMMENFASSUNG
We demonstrated the effect of nortriptyline, a tricyclic antidepressant drug and serotonin reuptake inhibitor, on voltage-dependent K