Merck
Non-genotoxic activation of p53 through the RPL11-dependent ribosomal stress pathway.
Carcinogenesis (2014-10-27)
Lucia Morgado-Palacin, Susana Llanos, Manuel Urbano-Cuadrado, Carmen Blanco-Aparicio, Diego Megias, Joaquín Pastor, Manuel Serrano
ZUSAMMENFASSUNG
Nucleolar disruption has recently emerged as a relevant means to activate p53 through inhibition of HDM2 by ribosome-free RPL11. Most drugs that induce nucleolar disruption also possess important genotoxic activity, which can have lasting mutagenic effects. Therefore, it is of interest to identify compounds that selectively produce nucleolar disruption in the absence of DNA damage. Here, we have performed a high-throughput screening to search for nucleolar disruptors. We have identified an acridine derivative (PubChem CID-765471) previously known for its capacity to activate p53 independently of DNA damage, although the molecular mechanism underlying p53 activation had remained uncharacterized. We report that CID-765471 produces nucleolar disruption by inhibiting ribosomal DNA transcription in a process that includes the selective degradation of the RPA194 subunit of RNA polymerase I. Following nucleolar disruption, CID-765471 activates p53 through the RPL11/HDM2 pathway in the absence of detectable DNA damage. In a secondary screening of compounds approved for medical use, we identify two additional acridine derivatives, aminacrine and ethacridine, that operate in a similar manner as CID-765471. These findings provide the basis for non-genotoxic chemotherapeutic approaches that selectively target the nucleolus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Cycloheximid, ≥90% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Hydroxyharnstoff, 98%, powder
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Actidion, Biotechnology Performance Certified
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)