Merck
  • Stimulation of in-vitro angiogenesis by low concentrations of risedronate is mitigated by 1,25-dihydroxyvitamin D3 or 24,25-dihydroxyvitamin D3.
Stimulation of in-vitro angiogenesis by low concentrations of risedronate is mitigated by 1,25-dihydroxyvitamin D3 or 24,25-dihydroxyvitamin D3.
The Journal of steroid biochemistry and molecular biology (2014-12-03)
José Manuel Quesada-Gómez, Raquel Santiago-Mora, Cristina Navarro-Valverde, Gabriel Dorado, Antonio Casado-Díaz
ZUSAMMENFASSUNG
The nitrogen-containing or nitrogenous bisphosphonates (N-BF) are currently the main class of drugs used for the treatment of diseases characterized by an increased bone resorption. Preliminary data suggest that N-BF have also direct or indirect anti-tumoral effects, and recent evidence suggests that part of the anti-tumoral activity of N-BF may be attributed to their anti-angiogenic capacity when they are used at high concentrations. On the other hand, an optimal vitamin-D status seems to be necessary to maximize the bone response to N-BF. Our aim has been to evaluate the effect of risedronate, alone or in combination with either 1,25(OH)2D3 or 24,25(OH)2D3 (two main vitamin-D metabolites) on parameters related to the angiogenic capacity of human umbilical-vein endothelial cells (HUVEC). The studies of tube formation through in-vitro angiogenesis assays with Matrigel, chemotaxis and migration in a scratch assay showed that low concentrations of risedronate (0.01 to 1μM) stimulated angiogenesis and cellular migration in vitro. The presence of 1,25(OH)2D3 in the medium inhibited tubular-structure formation and cellular migration. In addition, the presence of 1,25 or 24,25(OH)2D3 in the culture medium also decreased the pro-angiogenic effects of low-concentrations of risedronate. These data show the differential effects of different concentrations of vitamin-D metabolites and risedronate on angiogenesis, thus stressing the importance of an adequate vitamin D status during medical treatment with risedronate. This article is part of a Special Issue entitled '17th Vitamin D Workshop'.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
In-vitro-Toxikologieassay-Kit, auf Basis von Resazurin