- Investigation of the inclusion of the herbicide metobromuron in native cyclodextrins by powder X-ray diffraction and isothermal titration calorimetry.
Investigation of the inclusion of the herbicide metobromuron in native cyclodextrins by powder X-ray diffraction and isothermal titration calorimetry.
Carbohydrate research (2009-10-06)
Vincent J Smith, Natalia M Rougier, Rita H de Rossi, Mino R Caira, Elba I Buján, Mariana A Fernández, Susan A Bourne
PMID19800052
ZUSAMMENFASSUNG
We report the formation of inclusion complexes between the phenylurea herbicide metobromuron [3-(p-bromophenyl)-1-methoxy-1-methylurea] and beta- and gamma-cyclodextrin in the solid state. Formation of crystalline inclusion complexes by the kneading method was confirmed by powder X-ray diffraction and further structural characterization using the principles of isostructurality followed. In addition, DeltaH degrees , DeltaS degrees , DeltaG degrees and the association constants (K) at 298K were determined for complex formation in solution using isothermal titration calorimetry. The magnitudes of K for the formation of 1:1 complexes between metobromuron and alpha-, beta- and gamma-CD were estimated as 598, 310 and 114, respectively.