Merck
Morphological tuning, self-assembly and optical properties of indium oxide nanocrystals.
Physical chemistry chemical physics : PCCP (2010-09-08)
Enyi Ye, Shuang-Yuan Zhang, Suo Hon Lim, Shuhua Liu, Ming-Yong Han
ZUSAMMENFASSUNG
In this paper, weak acids, weak bases or their mixtures were used as reaction media/coordinating ligands to achieve systematic morphological control over amphoteric indium oxide nanostructures. Different indium/oleic acid molar ratios from 1 : 0, 1 : 1, 1 : 2, 1 : 3, 1 : 6 and 1 : 15 in non-coordinating, weakly coordinating, strongly coordinating and their mixed media were adopted to prepare irregular aggregated nanoparticles and uniform regular/truncated octahedra, etc. In addition to their strong size-dependent absorption, single-crystalline indium oxide octahedra also gave a strong band-edge emission while irregular indium oxide aggregated nanoparticles only exhibited a weak deep-trap emission. Meanwhile, the truncated octahedra were self-assembled into either zigzag lines or pentagram patterns, and the regular octahedra and truncated cubes were self-assembled into hexagonally packed nanocrystal arrays. In addition, the formation mechanism of the various nanostructures under different conditions was investigated in detail.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trioctylamin, 98%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge