Merck
Relationship between carnitine, fatty acids and insulin resistance.
Gynakologisch-geburtshilfliche Rundschau (2009-01-01)
Alfred Lohninger, U Radler, S Jinniate, S Lohninger, H Karlic, S Lechner, D Mascher, A Tammaa, H Salzer
ZUSAMMENFASSUNG
Increased plasma free fatty acid (FFA) levels are a feature of insulin resistance and type 2 diabetes. The aim of the present study was to assess the effect of L-carnitine supplementation on plasma lipids and the expression of enzymes in peripheral mononucleated cells (PMNC) involved in the regulation of fatty acid and glucose oxidation. L-Carnitine supplementation of 2 g/day resulted in a significant decrease in plasma FFA and in a less pronounced diminution of the plasma triacylglycerols. In addition, a concomitant increase in the relative mRNA abundances of carnitine acyltransferases (5- to 10-fold) and of the carnitine carrier OCTN2 (12-fold) in PMNC of pregnant women was found. The results of the present study provide evidence that L-carnitine supplementation in pregnancy (2 g/day) avoids a striking increase in plasma FFA, which are thought to be the main cause of insulin resistance and consequently gestational diabetes mellitus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carnitin-Acetyltransferase aus Taubenbrustmuskel, ammonium sulfate suspension, ≥50 units/mg protein