Merck
Histone Deacetylase Inhibition Attenuates Aortic Remodeling in Rats under Pressure Overload.
BioMed research international (2020-08-11)
Hanna Jung, Eunjo Lee, Inkyeom Kim, Gun Jik Kim
ZUSAMMENFASSUNG
The use of histone deacetylase (HDAC) inhibitor is a novel therapeutic strategy for cardiovascular disease. Studies have shown that many HDAC inhibitors have the ability to reduce the aortic remodeling in various animal models. We hypothesized that the HDAC inhibitor, MGCD0103 (MGCD), attenuates aortic remodeling in rats under pressure overload-induced by transverse aortic constriction (TAC). The aortic ring tension analysis was conducted using the thoracic aorta. Sections of the aorta were visualized after hematoxylin and eosin, trichrome, and Verhoeff-van Gieson staining, and immunohistochemistry. The expression of genes related to aortic remodeling (αSMA, Mmp2, and Mmp9) and angiotensin receptors (Agtr1 and Agtr2) was determined by quantitative real-time polymerase chain reaction. There was a significant decrease in relaxation of the aorta when treated with MGCD. Fibrosis of the aortic wall and expression of angiotensin receptors increased in TAC rats, which was attenuated by MGCD. These results indicate that MGCD, an HDAC inhibitor, attenuates aortic remodeling in rats with TAC-induced pressure overload rats and may serve as a potential therapeutic target of antiaortic remodeling in pressure overload-induced hypertension-related diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MGCD0103, ≥95% (HPLC)