Merck
  • Identification of Phytoconstituents in Leea indica (Burm. F.) Merr. Leaves by High Performance Liquid Chromatography Micro Time-of-Flight Mass Spectrometry.
Identification of Phytoconstituents in Leea indica (Burm. F.) Merr. Leaves by High Performance Liquid Chromatography Micro Time-of-Flight Mass Spectrometry.
Molecules (Basel, Switzerland) (2019-02-20)
Deepika Singh, Yin-Yin Siew, Teck-Ian Chong, Hui-Chuing Yew, Samuel Shan-Wei Ho, Claire Sophie En-Shen Lim, Wei-Xun Tan, Soek-Ying Neo, Hwee-Ling Koh
ZUSAMMENFASSUNG
Leea indica (Vitaceae) is a Southeast Asian medicinal plant. In this study, an ethyl acetate fraction of L. indica leaves was studied for its phytoconstituents using high-performance liquid chromatography-electrospray ionization-mass spectrometry (HPLC-ESI-microTOF-Q-MS/MS) analysis. A total of 31 compounds of different classes, including benzoic acid derivatives, phenolics, flavonoids, catechins, dihydrochalcones, coumarins, megastigmanes, and oxylipins were identified using LC-MS/MS. Among them, six compounds including gallic acid, methyl gallate, (-)-epigallocatechin-3-O-gallate, myricetin-3-O-rhamnoside, quercetin-3-O-rhamnoside, and 4',6'-dihydroxy-4-methoxydihydrochalcone 2'-O-β-d-glucopyranoside were isolated and identified by NMR analysis. The LC-MS/MS analysis led to the tentative identification of three novel dihydrochalcones namely 4',6'-dihydroxy-4-methoxydihydrochalcone 2'-O-rutinoside, 4',6'-dihydroxy-4-methoxydihydrochalcone 2'-O-glucosylpentoside and 4',6'-dihydroxy-4-methoxydihydrochalcone 2'-O-(3″-O-galloyl)-β-d-glucopyranoside. The structural identification of novel dihydrochalcones was based on the basic skeleton of the isolated dihydrochalcone, 4',6'-dihydroxy-4-methoxydihydrochalcone 2'-O-β-d-glucopyranoside and characteristic LC-MS/MS fragmentation patterns. This is the first comprehensive analysis for the identification of compounds from L. indica using LC-MS. A total 24 compounds including three new dihydrochalcones were identified for the first time from the genus Leea.