Merck
CCAT1 promotes triple-negative breast cancer progression by suppressing miR-218/ZFX signaling.
Aging (2019-07-17)
Chunyong Han, Xuebiao Li, Qian Fan, Guangshu Liu, Jian Yin
ZUSAMMENFASSUNG
Long non-coding RNAs (lncRNAs) regulate cancer development and progression. Here, we investigated the role of the lncRNA CCAT1 in triple-negative breast cancer (TNBC). CCAT1 expression was higher in TNBC cells than normal breast epithelial cells. Additionally, CCAT1 expression was higher in TNBC patient tumor tissue than adjacent normal breast tissue. Silencing CCAT1 inhibited TNBC cell proliferation, migration, and invasion in vitro, and tumor growth and progression in vivo. Bioinformatics analysis revealed that microRNA-218 (miR-218) is a potential target of CCAT1. Silencing CCAT1 resulted in an increase in miR-218 expression and inhibited TNBC cell proliferation, migration, and invasion. Silencing miR-218 reversed the effects of CCAT1 knockdown on cell proliferation, migration, and invasion, suggesting that CCAT1 promotes TNBC progression by downregulating miR-218 expression. We identified the zinc finger protein ZFX as a putative downstream target of miR-218 through bioinformatics analysis. ZFX expression was higher in TNBC than normal breast cell lines and higher in TNBC tumor tissue than adjacent normal breast tissue. Overexpression of ZFX reversed the tumor-suppressive effects of miR-218 on TNBC cell proliferation, migration, and invasion. Our data indicate that CCAT1 promotes TNBC progression by targeting the miR-218/ZFX axis.