Merck
EP5427718001
Eppendorf® Rotor Lid
for F-45-30-11, coated
About This Item
UNSPSC-Code:
41103913
NACRES:
NB.82
Leistungsmerkmale
for F-45-30-11, coated
Hersteller/Markenname
5427718001
Eppendorf® 5427718001
Eppendorf®
Methode(n)
centrifugation: suitable
Kompatibilität
for use with 5430 R
for use with 5430
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG