Merck
69864
pET-28a(+) DNA - Novagen
Novagen′s pET-28a-c(+) vectors carry an N-terminal His•Tag/thrombin/T7•Tag configuration plus an optional C-terminal His•Tag sequence.
Synonym(e):
cloning vector, expression vector, pET Plasmid, vector
About This Item
UNSPSC-Code:
41106610