TEM-HC200CU100
Holey Carbon Supported Copper Grids
size 200 mesh, size 100 nm , Hole, box of 25
Synonym(e):
Holey Carbon - Cu, 200 mesh, 100 micron
Empfohlene Produkte
Materialien
carbon film (holey)
copper grid
Verpackung
box of 25
Größe
100 nm , Hole
200 mesh
Allgemeine Beschreibung
Holey carbon film supported copper TEM gird: hole size 100 nm on 300 mesh copper grid.