Merck
SYNA341922
Synthware twin hose connecting adapter
joint: ST/NS 19/22
Synonym(e):
Synthware A341922, Adapter, twin connecting hose
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A341922
Verwandte Kategorien
Allgemeine Beschreibung
Adapter has two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.