Merck
SLWSST108K
PYREX(R) SPARE PTFE KEYS FOR PYREX(R) STOPCOCKS
(straight key bore 8.0 MM)
Synonym(e):
Pyrex, SciLabware SST108K
Größe auswählen
1 EA
CHF 73.10
CHF 73.10
Voraussichtliches Versanddatum15. April 2025
Größe auswählen
Ansicht ändern
1 EA
CHF 73.10
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.43
CHF 73.10
Voraussichtliches Versanddatum15. April 2025
Leistungsmerkmale
(straight key bore 8.0 MM)
Verpackung
pkg of 5 ea
Hersteller/Markenname
SciLabware SST108K
SciLabware
Allgemeine Beschreibung
Stopcocks, plain arms, straight bore, PTFE key
  • Manufactured from borosilicate glass with PTFE keys for highest resistance to chemical attack
  • Barrels are highly polished and the keys are self lubricating, avoiding the need for stopcock grease
  • Keys are fully interchangeable and supplied complete with plastic screw thread retaining device
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)