GF38258264
Inconel® 625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5
foil, 150 x 150mm, 0.2mm thickness, annealed
Synonym(e):
NI040240
Empfohlene Produkte
Form
foil (annealed)
Hersteller/Markenname
Goodfellow 382-582-64
Größe × Dicke
150 mm × 150 mm × 0.2 mm
Verwandte Kategorien
Allgemeine Beschreibung
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Rechtliche Hinweise
INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation