GC Analysis of Aromatics in Reformulated Gasoline on TCEP
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
TCEP, 60 m x 0.25 mm I.D., 0.44 μm (24153)
oven
50 °C (3 min), 10 °C/min to 140 °C (35 min)
inj. temp.
220 °C
detector
FID, 170 °C
carrier gas
helium, 30 cm/sec constant
injection
1 μL, 100:1 split
liner
4 mm I.D. FocusLiner™ inlet liner with taper
sample
Reformulated gasoline
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.