Merck
  • Cytoprotection by omega-3 fatty acids as a therapeutic drug vehicle when combined with nephrotoxic drugs in an intravenous emulsion: Effects on intraglomerular mesangial cells.
Cytoprotection by omega-3 fatty acids as a therapeutic drug vehicle when combined with nephrotoxic drugs in an intravenous emulsion: Effects on intraglomerular mesangial cells.
Toxicology reports (2014-10-22)
Gabriel Alejandro Bonaterra, Florian Wakenhut, Doris Röthlein, Martin Wolf, Bruce Ryan Bistrian, David Driscoll, Ralf Kinscherf
ABSTRACT
During therapeutic interventions, blood concentrations of intravenously applied drugs are higher, and their onset of pharmacological action is faster than with other routes of drug administration. However, acute drug therapy often produces nephrotoxic side effects, as commonly seen after treatment with Ketorolac or Gentamicin leading to questions about their use, especially for patients at risk for acute renal failure. Omega-6(n-6) and omega-3(n-3) polyunsaturated fatty acids (PUFA) affect eicosanoid metabolism, which plays a role in the regulation of inflammation. Eicosanoids derived from n-6 FA have proinflammatory and immunoactive functions, whereas eicosanoids derived from n-3 PUFA have anti-inflammatory and cytoprotective properties. We hypothesized that providing such injectable drugs with nephrotoxic potential in combination with n3-PUFAs from the outset, might afford rapid cytoprotection of renal cells, given the recent evidence that intravenously administered n3-PUFAs are rapidly incorporated into cell membranes. We used intraglomerular mesangial cells (MES13) that are sensitive to treatment with Ketorolac or Gentamicin instead of proximal tubular cells which do not respond to Ketorolac. We found a significant inhibition of Ketorolac (0.25, 0.5, 1 mM) or Gentamicin (2.5, 5 mM) induced cytotoxicity after pretreatment of MES13 cells with 0.01% of 20%w/v LipOmega-3 Emulsion 9/1, containing 90:10 wt/wt mixture of fish oil derived triglycerides to medium chain triglycerides.
Sigma-Aldrich
Anti-p62/SQSTM1 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-ATG5 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody