Merck
  • Therapeutic Targeting of the CBP/p300 Bromodomain Blocks the Growth of Castration-Resistant Prostate Cancer.
Therapeutic Targeting of the CBP/p300 Bromodomain Blocks the Growth of Castration-Resistant Prostate Cancer.
Cancer research (2017-08-19)
Lingyan Jin, Jesse Garcia, Emily Chan, Cecile de la Cruz, Ehud Segal, Mark Merchant, Samir Kharbanda, Ryan Raisner, Peter M Haverty, Zora Modrusan, Justin Ly, Edna Choo, Susan Kaufman, Maureen H Beresini, F Anthony Romero, Steven Magnuson, Karen E Gascoigne
ABSTRACT
Resistance invariably develops to antiandrogen therapies used to treat newly diagnosed prostate cancers, but effective treatments for castration-resistant disease remain elusive. Here, we report that the transcriptional coactivator CBP/p300 is required to maintain the growth of castration-resistant prostate cancer. To exploit this vulnerability, we developed a novel small-molecule inhibitor of the CBP/p300 bromodomain that blocks prostate cancer growth
Sigma-Aldrich
R1881, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Pricing and availability is not currently available.