Merck
  • Regulation of antimicrobial activity and xenocoumacins biosynthesis by pH in Xenorhabdus nematophila.
Regulation of antimicrobial activity and xenocoumacins biosynthesis by pH in Xenorhabdus nematophila.
Microbial cell factories (2017-11-17)
Shuqi Guo, Shujing Zhang, Xiangling Fang, Qi Liu, Jiangtao Gao, Muhammad Bilal, Yonghong Wang, Xing Zhang
ABSTRACT
Xenocoumacin 1 (Xcn1) and Xenocoumacin 2 (Xcn2) are the main antimicrobial compounds produced by Xenorhabdus nematophila. Culture conditions, including pH, had remarkably distinct effects on the antimicrobial activity of X. nematophila. However, the regulatory mechanism of pH on the antimicrobial activity and antibiotic production of this bacterium is still lacking. With the increase of initial pH, the antimicrobial activity of X. nematophila YL001 was improved. The levels of Xcn1 and nematophin at pH 8.5 were significantly (P < 0.05) higher than that at pH 5.5 and 7.0. In addition, the expression of xcnA-L, which are responsible for the production of Xcn1 was increased and the expression of xcnMN, which are required for the conversion of Xcn1 to Xcn2 was reduced at pH 8.5. Also, the expression of ompR and cpxR were decreased at pH 8.5. The alkaline pH environment was found to be beneficial for the production of Xcn1 and nematophin, which in turn led to high antimicrobial activity of X. nematophila at pH 8.5.
Product Description
Supelco
Amberlite XAD16N, 20-60 mesh
Pricing and availability is not currently available.