Merck
  • Cyclooxygenase 2 is a key enzyme for inflammatory cytokine-induced angiogenesis.
Cyclooxygenase 2 is a key enzyme for inflammatory cytokine-induced angiogenesis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2004-02-11)
Takashi Kuwano, Shintaro Nakao, Hidetaka Yamamoto, Masazumi Tsuneyoshi, Tomoya Yamamoto, Michihiko Kuwano, Mayumi Ono
ABSTRACT
Cyclooxygenase1 (COX1) and COX2 mediate the rate-limiting step in arachidonic acid metabolism. Expression of COX2 mRNA and protein is often enhanced in various human cell types by inflammatory cytokines such as interleukin-1beta (IL-1beta) and tumor necrosis factor alpha (TNFalpha). IL-1beta enhanced expression of various prostanoids and this expression was blocked by COX2 selective inhibitors. IL-1beta markedly induced angiogenesis in vitro and in vivo, which was significantly inhibited by COX2 selective inhibitors but not by a vascular endothelial growth factor (VEGF) receptor tyrosine kinase inhibitor. In contrast, COX2 selective inhibitors only partially blocked VEGF-induced angiogenesis. EP2, EP4 (prostaglandin E2 receptors) agonists and thromboxane A2 (TXA2) receptor agonists induced angiogenesis in vitro and in vivo; IL-1beta-induced angiogenesis was blocked by an EP4 antagonist and a TXA2 receptor antagonist. IL-1beta induced much less angiogenesis in cornea of COX2 knockout mice than that of wild-type mice. This is the first report that COX2 and some prostanoids play a key role in IL-1beta-induced angiogenesis.
Sigma-Aldrich
ONO-AE3-208, ≥98% (HPLC)
Pricing and availability is not currently available. Contact Technical Service