Merck
  • Microwave-Assisted Synthesis of Heteroleptic Ir(III)(+) Polypyridyl Complexes.
Microwave-Assisted Synthesis of Heteroleptic Ir(III)(+) Polypyridyl Complexes.
The Journal of organic chemistry (2016-06-16)
Timothy M Monos, Alexandra C Sun, Rory C McAtee, James J Devery, Corey R J Stephenson
ABSTRACT
We report a rapid, one-pot, operationally simple, and scalable preparation of valuable cationic heteroleptic iridium(III) polypyridyl photosensitizers. This method takes advantage of two consecutive microwave irradiation steps in the same reactor vial, avoiding the need for additional reaction purifications. A number of known heteroleptic iridium(III) complexes are prepared in up to 96% yield. Notably, this method is demonstrated to provide the synthetically versatile photosensitizer [Ir(ppy)2(dtbbpy)]PF6 in >1 g quantities in less than 5 h of bench time. We envision this method will help accelerate future developments in visible-light-dependent chemistry.
Sigma-Aldrich
[Ir(dFppy)2(dtbbpy)]PF6
Pricing and availability is not currently available.