Merck
  • Reactive Precipitation of Anhydrous Alkali Sulfide Nanocrystals with Concomitant Abatement of Hydrogen Sulfide and Cogeneration of Hydrogen.
Reactive Precipitation of Anhydrous Alkali Sulfide Nanocrystals with Concomitant Abatement of Hydrogen Sulfide and Cogeneration of Hydrogen.
ChemSusChem (2017-05-17)
Xuemin Li, Yangzhi Zhao, Alice Brennan, Miranda McCeig, Colin A Wolden, Yongan Yang
ABSTRACT
Anhydrous alkali sulfide (M
Sigma-Aldrich
2-Methyltetrahydrofuran, BioRenewable, anhydrous, ≥99%, Inhibitor-free
Sigma-Aldrich
Benzene-D6, anhydrous, ≥99.6 atom % D
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
Dibutyl ether, anhydrous, 99.3%