Merck
Correlated optical and isotopic nanoscopy.
Nature communications (2014-04-11)
Sinem K Saka, Angela Vogts, Katharina Kröhnert, François Hillion, Silvio O Rizzoli, Johannes T Wessels
ABSTRACT
The isotopic composition of different materials can be imaged by secondary ion mass spectrometry. In biology, this method is mainly used to study cellular metabolism and turnover, by pulsing the cells with marker molecules such as amino acids labelled with stable isotopes ((15)N, (13)C). The incorporation of the markers is then imaged with a lateral resolution that can surpass 100 nm. However, secondary ion mass spectrometry cannot identify specific subcellular structures like organelles, and needs to be correlated with a second technique, such as fluorescence imaging. Here, we present a method based on stimulated emission depletion microscopy that provides correlated optical and isotopic nanoscopy (COIN) images. We use this approach to study the protein turnover in different organelles from cultured hippocampal neurons. Correlated optical and isotopic nanoscopy can be applied to a variety of biological samples, and should therefore enable the investigation of the isotopic composition of many organelles and subcellular structures.
Sigma-Aldrich
Monoclonal Anti-TOMM20 antibody produced in mouse, clone 4F3, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.