Merck
  • Pentraxin-3 serum levels are associated with disease severity and mortality in patients with systemic inflammatory response syndrome.
Pentraxin-3 serum levels are associated with disease severity and mortality in patients with systemic inflammatory response syndrome.
PloS one (2013-09-17)
Simone Bastrup-Birk, Mikkel-Ole Skjoedt, Lea Munthe-Fog, Jens J Strom, Ying Jie Ma, Peter Garred
ABSTRACT
The long pentraxin-3 (PTX3) is a key component of the humoral arm of the innate immune system. PTX3 is produced locally in response to pro-inflammatory stimuli. To investigate PTX3 levels and its use as a biomarker in patients with systemic inflammation, we developed a solid-phase enzyme-linked immunosorbent assay based on novel anti-PTX3 monoclonal antibodies detecting PTX3 with high sensitivity. The assay was applied on 261 consecutive patients admitted to an intensive care unit prospectively monitored with the systemic inflammatory response syndrome (SIRS). 100 blood donors were included as controls. PTX3 levels were elevated in patients (median = 71.3 ng/ml) compared with the controls (median = 0 ng/ml) (Mann-Whitney, p<0.0001). ROC analysis showed that PTX3 levels were significantly specific (85.0%) and sensitive (89.1%) to discriminate between healthy controls and patients (area under the curve (AUC) 0.922 (95% CI 0.892 to 0.946, p<0.0001)). Higher levels of PTX3 were associated with the development of sepsis, severe sepsis and septic shock (p = 0.0001). The serum levels of PTX3 correlated significantly with SAPS2 score (Spearman's rho 0.28, p<0.0001). Patients with high levels of PTX3 at admission did have a higher 90 day mortality rate than patients with the 25% lowest levels (Cox regression analysis, hazard ratio 3.0, p = 0.0009). In conclusion, we have established a highly sensitive and robust assay for measurement of PTX3 and found that its serum concentrations correlated with disease severity and mortality in patients with SIRS and sepsis.
Sigma-Aldrich
Pentraxin 3 human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Pricing and availability is not currently available.