Merck
  • MLi-2, a Potent, Selective, and Centrally Active Compound for Exploring the Therapeutic Potential and Safety of LRRK2 Kinase Inhibition.
MLi-2, a Potent, Selective, and Centrally Active Compound for Exploring the Therapeutic Potential and Safety of LRRK2 Kinase Inhibition.
The Journal of pharmacology and experimental therapeutics (2015-09-27)
Matthew J Fell, Christian Mirescu, Kallol Basu, Boonlert Cheewatrakoolpong, Duane E DeMong, J Michael Ellis, Lynn A Hyde, Yinghui Lin, Carrie G Markgraf, Hong Mei, Michael Miller, Frederique M Poulet, Jack D Scott, Michelle D Smith, Zhizhang Yin, Xiaoping Zhou, Eric M Parker, Matthew E Kennedy, John A Morrow
ABSTRACT
Mutations in the leucine-rich repeat kinase 2 (LRRK2) gene are the most common genetic cause of familial and sporadic Parkinson's disease (PD). That the most prevalent mutation, G2019S, leads to increased kinase activity has led to a concerted effort to identify LRRK2 kinase inhibitors as a potential disease-modifying therapy for PD. An internal medicinal chemistry effort identified several potent and highly selective compounds with favorable drug-like properties. Here, we characterize the pharmacological properties of cis-2,6-dimethyl-4-(6-(5-(1-methylcyclopropoxy)-1H-indazol-3-yl)pyrimidin-4-yl)morpholine (MLi-2), a structurally novel, highly potent, and selective LRRK2 kinase inhibitor with central nervous system activity. MLi-2 exhibits exceptional potency in a purified LRRK2 kinase assay in vitro (IC50 = 0.76 nM), a cellular assay monitoring dephosphorylation of LRRK2 pSer935 LRRK2 (IC50 = 1.4 nM), and a radioligand competition binding assay (IC50 = 3.4 nM). MLi-2 has greater than 295-fold selectivity for over 300 kinases in addition to a diverse panel of receptors and ion channels. Acute oral and subchronic dosing in MLi-2 mice resulted in dose-dependent central and peripheral target inhibition over a 24-hour period as measured by dephosphorylation of pSer935 LRRK2. Treatment of MitoPark mice with MLi-2 was well tolerated over a 15-week period at brain and plasma exposures >100× the in vivo plasma IC50 for LRRK2 kinase inhibition as measured by pSer935 dephosphorylation. Morphologic changes in the lung, consistent with enlarged type II pneumocytes, were observed in MLi-2-treated MitoPark mice. These data demonstrate the suitability of MLi-2 as a compound to explore LRRK2 biology in cellular and animal models.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone 2/40/15, clone 2/40/15, Chemicon®, from mouse
Sigma-Aldrich
MLi-2, ≥98% (HPLC)
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
DL-Tyrosine, 99%