Merck
  • Rates of serious infections and malignancies among patients with rheumatoid arthritis receiving either tumor necrosis factor inhibitor or rituximab therapy.
Rates of serious infections and malignancies among patients with rheumatoid arthritis receiving either tumor necrosis factor inhibitor or rituximab therapy.
The Journal of rheumatology (2015-01-17)
Kalle Jyri Aaltonen, Jaana Tuulikki Joensuu, Liisa Virkki, Tuulikki Sokka, Pasi Aronen, Heikki Relas, Heikki Valleala, Vappu Rantalaiho, Laura Pirilä, Kari Puolakka, Tea Uusitalo, Marja Blom, Yrjö Tapio Konttinen, Dan Nordström
ABSTRACT
Because of the role of tumor necrosis factor (TNF) in host defense, it was hypothesized that its inhibition might lead to an increased risk of malignancies and infections. The objective of our study was to assess the incidence of serious infections leading to hospitalization and malignancies among patients with rheumatoid arthritis (RA) receiving either TNF inhibitor or rituximab (RTX) therapy. The study population was identified from the National Register for Biologic Treatment in Finland and the hospital records of Central Finland Central Hospital for conventional disease-modifying antirheumatic drug (cDMARD) users. Data on infections and malignancies were acquired from national healthcare registers. A Poisson model was used to calculate the adjusted incidence rate ratios (aIRR) and was composed of age, sex, time from diagnosis, year of the beginning of the followup, rheumatoid factor status, Disease Activity Score at 28 joints, Health Assessment Questionnaire, prior malignancy, prior serious infection, prior biologic use, and time-updated use of methotrexate, sulfasalazine, hydroxychloroquine, and oral corticosteroids as confounders. In total, during the followup of 10,994 patient-years, 92 malignancies and 341 serious infections were included in the analyses. The aIRR of infections compared to cDMARD users were 1.2 (95% CI 0.63-2.3), 0.84 (95% CI 0.53-1.3), 0.98 (95% CI 0.60-1.6), and 1.1 (95% CI 0.59-1.9) for the patients treated with infliximab (IFX), etanercept, adalimumab, and RTX, respectively. The crude rates of malignancies were highest among the users of cDMARD and RTX, and lowest among patients treated with IFX with no differences in aIRR. Our results provide some reassurance of the safety of biologic treatments in the treatment of RA.
Product Description
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Methotrexate, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cortisone, ≥95%