Merck
  • Tumours of histiocytes and accessory dendritic cells: an immunohistochemical approach to classification from the International Lymphoma Study Group based on 61 cases.
Tumours of histiocytes and accessory dendritic cells: an immunohistochemical approach to classification from the International Lymphoma Study Group based on 61 cases.
Histopathology (2002-07-18)
S A Pileri, T M Grogan, N L Harris, P Banks, E Campo, J K C Chan, R D Favera, G Delsol, C De Wolf-Peeters, B Falini, R D Gascoyne, P Gaulard, K C Gatter, P G Isaacson, E S Jaffe, P Kluin, D M Knowles, D Y Mason, S Mori, H-K Müller-Hermelink, M A Piris, E Ralfkiaer, H Stein, I-J Su, R A Warnke, L M Weiss
ABSTRACT
Neoplasms of histiocytes and dendritic cells are rare, and their phenotypic and biological definition is incomplete. Seeking to identify antigens detectable in paraffin-embedded sections that might allow a more complete, rational immunophenotypic classification of histiocytic/dendritic cell neoplasms, the International Lymphoma Study Group (ILSG) stained 61 tumours of suspected histiocytic/dendritic cell type with a panel of 15 antibodies including those reactive with histiocytes (CD68, lysozyme (LYS)), Langerhans cells (CD1a), follicular dendritic cells (FDC: CD21, CD35) and S100 protein. This analysis revealed that 57 cases (93%) fit into four major immunophenotypic groups (one histiocytic and three dendritic cell types) utilizing six markers: CD68, LYS, CD1a, S100, CD21, and CD35. The four (7%) unclassified cases were further classifiable into the above four groups using additional morphological and ultrastructural features. The four groups then included: (i) histiocytic sarcoma (n=18) with the following phenotype: CD68 (100%), LYS (94%), CD1a (0%), S100 (33%), CD21/35 (0%). The median age was 46 years. Presentation was predominantly extranodal (72%) with high mortality (58% dead of disease (DOD)). Three had systemic involvement consistent with 'malignant histiocytosis'; (ii) Langerhans cell tumour (LCT) (n=26) which expressed: CD68 (96%), LYS (42%), CD1a (100%), S100 (100%), CD21/35 (0%). There were two morphological variants: cytologically typical (n=17) designated LCT; and cytologically malignant (n=9) designated Langerhans cell sarcoma (LCS). The LCS were often not easily recognized morphologically as LC-derived, but were diagnosed based on CD1a staining. LCT and LCS differed in median age (33 versus 41 years), male:female ratio (3.7:1 versus 1:2), and death rate (31% versus 50% DOD). Four LCT patients had systemic involvement typical of Letterer-Siwe disease; (iii) follicular dendritic cell tumour/sarcoma (FDCT) (n=13) which expressed: CD68 (54%), LYS (8%), CD1a (0%), S100 (16%), FDC markers CD21/35 (100%), EMA (40%). These patients were adults (median age 65 years) with predominantly localized nodal disease (75%) and low mortality (9% DOD); (iv) interdigitating dendritic cell tumour/sarcoma (IDCT) (n=4) which expressed: CD68 (50%), LYS (25%), CD1a (0%), S100 (100%), CD21/35 (0%). The patients were adults (median 71 years) with localized nodal disease (75%) without mortality (0% DOD). In conclusion, definitive immunophenotypic classification of histiocytic and accessory cell neoplasms into four categories was possible in 93% of the cases using six antigens detected in paraffin-embedded sections. Exceptional cases (7%) were resolvable when added morphological and ultrastructural features were considered. We propose a classification combining immunophenotype and morphology with five categories, including Langerhans cell sarcoma. This simplified scheme is practical for everyday diagnostic use and should provide a framework for additional investigation of these unusual neoplasms.
Sigma-Aldrich
CD21 (EP3093) Rabbit Monoclonal Primary Antibody