Merck
  • Lower homologues of ahpatinin, aspartic protease inhibitors, from a marine Streptomyces sp.
Lower homologues of ahpatinin, aspartic protease inhibitors, from a marine Streptomyces sp.
Journal of natural products (2014-06-25)
Yi Sun, Kentaro Takada, Yuichi Nogi, Shigeru Okada, Shigeki Matsunaga
ABSTRACT
Two linear peptides, ahpatinin Ac (1) and ahpatinin Pr (2), were isolated together with the known ahpatinin (i)Bu, pepstatin Ac, pepstatin Pr, and pepsinostreptin from a Streptomyces sp. derived from a deep-sea sediment. The structure of ahpatinin Pr (2) was assigned by interpretation of NMR data and HPLC analysis of the hydrolysate after converting to the DNP-L-Val derivative. During the LCMS analysis of the acid hydrolysate, products arising from the retro-aldol cleavage of the statine and Ahppa units in 2 were observed and could facilitate the determination of the absolute configuration of the statine class of nonproteinogenic amino acids. Both ahpatinin Ac (1) and ahpatinin Pr (2) potently inhibited pepsin and moderately inhibited cathepsin B.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥3200 units/mg protein