Merck
  • Intracerebral hemorrhage after lumbar myelography with iohexol: report of a case and review of the literature.
Intracerebral hemorrhage after lumbar myelography with iohexol: report of a case and review of the literature.
Neurosurgery (1991-04-01)
E Van de Kelft, J Bosmans, P M Parizel, M Van Vyve, P Selosse
ABSTRACT
Intracranial hemorrhage is an uncommon complication of dural puncture. In most instances, hematomas are subdural; they may be unilateral or bilateral. Rarely are intraparenchymal cerebral hemorrhages related to dural puncture. This report describes a delayed occurrence of bilateral intraparenchymal hemorrhages in a 38-year-old woman 7 days after lumbar myelography with iohexol. A review of the literature is presented.
Product Description
Iohexol for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Pricing and availability is not currently available.
Iohexol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Iohexol, analytical standard
Pricing and availability is not currently available.