Merck
  • Vitamin D signaling pathway plays an important role in the development of heart failure after myocardial infarction.
Vitamin D signaling pathway plays an important role in the development of heart failure after myocardial infarction.
Journal of applied physiology (Bethesda, Md. : 1985) (2013-02-23)
Soochan Bae, Sylvia S Singh, Hyeon Yu, Ji Yoo Lee, Byung Ryul Cho, Peter M Kang
ABSTRACT
Accumulating evidence suggests that vitamin D deficiency plays a crucial role in heart failure. However, whether vitamin D signaling itself plays an important role in cardioprotection is poorly understood. In this study, we examined the mechanism of modulating vitamin D signaling on progression to heart failure after myocardial infarction (MI) in mice. Vitamin D signaling was activated by administration of paricalcitol (PC), an activated vitamin D analog. Wild-type (WT) mice underwent sham or MI surgery and then were treated with either vehicle or PC. Compared with vehicle group, PC attenuated development of heart failure after MI associated with decreases in biomarkers, apoptosis, inflammation, and fibrosis. There was also improvement of cardiac function with PC treatment after MI. Furthermore, vitamin D receptor (VDR) mRNA and protein levels were restored by PC treatment. Next, to explore whether defective vitamin D signaling exhibited deleterious responses after MI, WT and VDR knockout (KO) mice underwent sham or MI surgery and were analyzed 4 wk after MI. VDR KO mice displayed a significant decline in survival rate and cardiac function compared with WT mice after MI. VDR KO mice also demonstrated a significant increase in heart failure biomarkers, apoptosis, inflammation, and fibrosis. Vitamin D signaling promotes cardioprotection after MI through anti-inflammatory, antifibrotic and antiapoptotic mechanisms.
Sigma-Aldrich
Paricalcitol, ≥98% (HPLC)
USP
Paricalcitol, United States Pharmacopeia (USP) Reference Standard