Merck
  • In vivo anti-cancer efficacy of magnetite nanocrystal--based system using locoregional hyperthermia combined with 5-fluorouracil chemotherapy.
In vivo anti-cancer efficacy of magnetite nanocrystal--based system using locoregional hyperthermia combined with 5-fluorouracil chemotherapy.
Biomaterials (2013-07-24)
Tsung-Ju Li, Chih-Chia Huang, Pin-Wei Ruan, Kuei-Yi Chuang, Kuang-Jing Huang, Dar-Bin Shieh, Chen-Sheng Yeh
ABSTRACT
We present an approach for synchronizing hyperthermia and thermal-responsive local drug release. The targeting probe has a magnetite nanocrystal (Fe₃O₄@PSMA) core and a polynucleotide shell that carries 5-fluorouracil (5-FU) and anti-human epidermal growth factor receptor 2 (anti-HER2) antibody for cancer cell-specific targeting. The targeting nanocrystals play as an important role to relay the externally delivered radiofrequency energy for tumor hyperthermia. Locoregional heat then triggers a drug release from the oligonucleotide carrier as it directly damages tumor cells. Cell viability assays and pathological examinations show that this synchronization is significantly more efficacious in both in vitro and in vivo models than hyperthermia or chemotherapy alone. Prominent tumor remission in vivo was achieved through radiofrequency synchronization of hyperthermia and chemotherapy after the nanoparticle had been intravenously injected.
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron(II,III) oxide, powder, <5 μm, 95%
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron(II,III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene