Merck
  • Salivary estriol concentrations during normal pregnancies, and a comparison with plasma estriol.
Salivary estriol concentrations during normal pregnancies, and a comparison with plasma estriol.
Clinical chemistry (1984-01-01)
J J Evans, A R Wilkinson, D R Aickin
ABSTRACT
Saliva would have advantages over plasma or urine for monitoring estriol during pregnancy. Specimen collection, after stimulation of flow by citric acid, is non-invasive and simple. We measured concentrations of unconjugated estriol in saliva and compared them with those in plasma in normal pregnancies, and found a good correlation (r = 0.79). In addition, trends of concentrations in saliva and plasma were statistically compared and found to be highly correlated. The variation among individuals in the saliva/plasma concentration ratio suggested that some inter-individual factor(s) may affect this relationship. The normal reference interval for unconjugated estriol concentration in saliva from 20 weeks of gestation to term was established.
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Estriol, meets USP testing specifications
Supelco
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®