Merck
  • [Postmortem detection of accidental methadone intoxication in a 10-year-old girl].
[Postmortem detection of accidental methadone intoxication in a 10-year-old girl].
Archiv fur Kriminologie (2013-07-25)
Annette Thierauf, Susanne Vogt, Stefan Pollak, Markus Grosse Perdekamp
ABSTRACT
Intoxications in children still account for a large part of the cases reported to poison control centres. Among drug intoxications, substitution drugs have gained in importance lately. Careless storage of take-home medications puts children living in the same household at risk. The authors report on a fatal case of a 10-year-old girl who died from intoxication with methadone belonging to a carer. The accidental intoxication had not been diagnosed at the hospital where the girl was treated for suspected meningitis. The question of medical malpractice is discussed.
Sigma-Aldrich
(±)-Methadone hydrochloride, powder, ≥98%
Supelco
(±)-Methadone solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®