Merck
  • Dentist's occupational allergic contact dermatitis caused by coconut diethanolamide, N-ethyl-4-toluene sulfonamide and 4-tolyldiethanolamine.
Dentist's occupational allergic contact dermatitis caused by coconut diethanolamide, N-ethyl-4-toluene sulfonamide and 4-tolyldiethanolamine.
Acta dermato-venereologica (1993-04-01)
L Kanerva, R Jolanki, T Estlander
ABSTRACT
Dental personnel are exposed to many sensitizing compounds at work and often develop multiple delayed allergies. Here we report on a dentist who got sensitized to several products that have not, or only seldom, caused sensitization earlier. These products were: coconut diethanolamide from her handwashing liquids, N-ethyl-4-toluene sulfonamide, a resin carrier in dental materials for isolating cavities underneath restorations, and 4-tolyldiethanolamine, an accelerator for inducing polymerization of dental acrylic resins at room temperature. The patient also had allergic patch test reactions to formaldehyde, phenol-formaldehyde resin, fragrance mix, and lauryl monoethanolamide, possibly from occupational exposure.
Sigma-Aldrich
2,2′-(4-Methylphenylimino)diethanol, ≥97.0% (NT)
Sigma-Aldrich
2,2′-(4-Methylphenylimino)diethanol, technical grade, 90%