Merck
  • Production and certification of an enzyme reference material for gamma-glutamyltransferase (CRM 319). Part 2: Certification campaign.
Production and certification of an enzyme reference material for gamma-glutamyltransferase (CRM 319). Part 2: Certification campaign.
Clinical chemistry (1987-11-01)
F Schiele, J Muller, E Colinet, G Siest
ABSTRACT
We describe the process of certification for a gamma-glutamyltransferase reference material (CRM no. 319). Fifteen laboratories participated to this interlaboratory evaluation. All steps of the measurements were controlled in an effort to locate potential sources of variations. In particular, the exclusion of some data was strictly documented or justified by the non-observance of the IFCC method and (or) discrepancies in instrumentation, reconstitution of the lyophilized samples, or measurement technique. Inaccuracy in the reconstitution of the lyophilized material was +/- 0.68%, and the molar absorptivity of the 5-amino-2-nitrobenzoate reported by each laboratory was within +/- 2% limits of the value reported by the IFCC. Calculated from the sets of accepted results, the total CV among samples was 2.6% and the overall CV was 3.2%. Within-day and between-day CVs were 1.1% and 1.4%, respectively. The greatest variation for a single component was the between-laboratory variability (CV 3.1%); the within-laboratory CV, including the day effect, was 1.8%. Finally, the certified value for the catalytic concentration of this enzyme in the reconstituted lyophilized reference material was 86.8 U/L with an uncertainty of +/- 2.1 U/L (0.95 confidence interval). The uncertainty appeared to be compatible with the end-use of this reference material.
Sigma-Aldrich
5-Amino-2-nitrobenzoic acid, 97%
Pricing and availability is not currently available.