Merck
  • [Study of glutamine solution use efficiency in pediatric patients with heavy thermic burns and concomitant injuries in the intensive care unit].
[Study of glutamine solution use efficiency in pediatric patients with heavy thermic burns and concomitant injuries in the intensive care unit].
Anesteziologiia i reanimatologiia (2013-07-03)
A U Lekmanov, U V Erpuleva, I V Zolkina, P A Rossaus
ABSTRACT
Comparing analysis of randomized study was conducted. 40 Pediatric patients (2-15 years old) with heavy thermic burns and concomitant injuries were recruited into the study to assess the glutamine solution intravenous use efficiency. Parenteral feeding and feeding with glutamine solution intravenous use were clinicaly and laboratory compared Duration of mechanical ventilation decreased in group of feeding with glutamine solution intravenous use. Preliminary results of the study recommend to include glutamine in the programs of parentreral feeding.
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material