Merck
  • Visible and near-infrared planar waveguide structure of polycrystalline zinc sulfide from C ions implantation.
Visible and near-infrared planar waveguide structure of polycrystalline zinc sulfide from C ions implantation.
Optics express (2013-03-14)
Tao Liu, Peng Liu, Lian Zhang, Yu-Fan Zhou, Xiao-Fei Yu, Jin-Hua Zhao, Xue-Lin Wang
ABSTRACT
We report the fabrication of a planar waveguide in polycrystalline zinc sulfide by 6.0 MeV C ions implantation with a fluence of 5 × 10¹⁴ ion/cm² at room temperature. The near-field light intensity profiles in the visible and near-infrared bands are measured by the end-face coupling method with different laser sources. Investigation of the Raman spectra demonstrates that the microstructure of the polycrystalline zinc sulfide has no significant change after C ion implantation. The absorption spectra show that the implantation processes have no influence on the visible and infrared bands.
Sigma-Aldrich
Zinc sulfide, powder, 10 μm, 99.99% trace metals basis
Sigma-Aldrich
Zinc sulfide, pieces, 3-12 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc sulfide, purum, 97% (from Zn)